Assay Details
Gene Symbol: ARPP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002753452.4 | XP_002753498.1 | 3 - 4 | 222 | 77 | |
XM_009005462.2 | XP_009003710.1 | 3 - 4 | 367 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
