Assay Details
| Gene Symbol: SVIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755119.4 | XP_002755165.1 | 3 - 4 | 336 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SVIP| Gene Symbol: SVIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755119.4 | XP_002755165.1 | 3 - 4 | 336 | 101 | |
Set Membership: |
|