Assay Details
| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758237.3 | XP_002758283.1 | 3 - 4 | 534 | 92 | |
| XM_008981773.2 | XP_008980021.1 | 3 - 4 | 534 | 92 | ||
More Information
Set Membership: |
|

TLR9
Genomic Map