Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759786.3 | XP_002759832.1 | 5 - 6 | 1018 | 74 | |
| XM_008984145.2 | XP_008982393.1 | 5 - 6 | 1048 | 74 | ||
| XM_008984146.2 | XP_008982394.1 | 4 - 5 | 764 | 74 | ||
More Information
Set Membership: |
|

ZNF713
Genomic Map