Assay Details
| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744370.3 | XP_002744416.1 | 3 - 4 | 448 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYOZ3| Gene Symbol: MYOZ3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744370.3 | XP_002744416.1 | 3 - 4 | 448 | 76 | |
Set Membership: |
|