Assay Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008991784.2 | XP_008990032.1 | 3 - 4 | 308 | 109 | |
| XM_008991786.2 | XP_008990034.1 | 3 - 4 | 780 | 109 | ||
| XM_017969778.1 | XP_017825267.1 | 4 - 5 | 403 | 109 | ||
| XM_017969779.1 | XP_017825268.1 | 3 - 4 | 345 | 109 | ||
| XM_017969781.1 | XP_017825270.1 | 4 - 5 | 1165 | 109 | ||
More Information
Set Membership: |
|

RFESD
Genomic Map