Assay Details
| Gene Symbol: LOC100398101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761942.4 | XP_002761988.2 | 5 - 6 | 869 | 94 | |
| XM_008987599.2 | XP_008985847.1 | 5 - 6 | 1624 | 94 | ||
More Information
Set Membership: |
|

LOC100398101
Genomic Map