Assay Details
| Gene Symbol: LOC100398101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761942.4 | XP_002761988.2 | 6 - 7 | 929 | 76 | |
| XM_008987599.2 | XP_008985847.1 | 6 - 7 | 1684 | 76 | ||
More Information
Set Membership: |
|

LOC100398101
Genomic Map