Assay Details
| Gene Symbol: OVOL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747470.3 | XP_002747516.1 | 3 - 4 | 1551 | 82 | |
| XM_008995802.2 | XP_008994050.1 | 4 - 5 | 793 | 82 | ||
| XM_017971977.1 | XP_017827466.1 | 4 - 5 | 803 | 82 | ||
| XM_017971978.1 | XP_017827467.1 | 2 - 3 | 227 | 82 | ||
More Information
Set Membership: |
|

OVOL2
Genomic Map