Assay Details
| Gene Symbol: KCNJ2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748668.3 | XP_002748714.1 | 1 - 2 | 184 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002748668.3 transcripts.
Set Membership: |
|

KCNJ2
Genomic Map