Assay Details
Gene Symbol: ZNF234 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008988174.2 | XP_008986422.1 | 5 | 1251 | 96 | |
XM_008988177.2 | XP_008986425.1 | 5 | 1248 | 96 | ||
XM_017967495.1 | XP_017822984.1 | 6 | 1347 | 96 | ||
XM_017967496.1 | XP_017822985.1 | 6 | 1357 | 96 | ||
XM_017967497.1 | XP_017822986.1 | 6 | 1344 | 96 | ||
XM_017967498.1 | XP_017822987.1 | 6 | 1354 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
