Assay Details
| Gene Symbol: PODXL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752001.2 | XP_002752047.2 | 2 - 3 | 983 | 70 | |
| XM_009003052.1 | XP_009001300.1 | 2 - 3 | 983 | 70 | ||
| XM_009003053.1 | XP_009001301.1 | 2 - 3 | 935 | 70 | ||
More Information
Set Membership: |
|

PODXL
Genomic Map