Assay Details
| Gene Symbol: SULT1C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757476.4 | XP_002757522.1 | 7 - 8 | 1020 | 68 | |
| XM_008980231.2 | XP_008978479.1 | 7 - 8 | 1028 | 68 | ||
More Information
Set Membership: |
|

SULT1C2
Genomic Map