Assay Details
| Gene Symbol: MGST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003276.2 | XP_009001524.1 | 2 - 3 | 259 | 70 | |
| XM_009003277.1 | XP_009001525.1 | 2 - 3 | 405 | 70 | ||
| XM_009003278.2 | XP_009001526.1 | 2 - 3 | 277 | 70 | ||
| XM_009003280.2 | XP_009001528.1 | 2 - 3 | 326 | 70 | ||
| XM_009003281.2 | XP_009001529.1 | 2 - 3 | 259 | 70 | ||
More Information
Set Membership: |
|

MGST1
Genomic Map