Assay Details
| Gene Symbol: TMPRSS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761447.2 | XP_002761493.1 | 12 - 13 | 1541 | 63 | |
| XM_008986736.1 | XP_008984984.1 | 12 - 13 | 2295 | 63 | ||
More Information
Set Membership: |
|

TMPRSS3
Genomic Map