Assay Details
| Gene Symbol: LDLRAD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979455.2 | XP_008977703.1 | 4 - 5 | 1778 | 81 | |
| XM_008979456.2 | XP_008977704.1 | 4 - 5 | 5565 | 81 | ||
| XM_008979457.2 | XP_008977705.1 | 4 - 5 | 2178 | 81 | ||
| XM_008979459.2 | XP_008977707.1 | 4 - 5 | 1003 | 81 | ||
| XM_008979463.2 | XP_008977711.1 | 2 - 3 | 1241 | 81 | ||
| XM_008979464.2 | XP_008977712.1 | 2 - 3 | 595 | 81 | ||
| XM_017963850.1 | XP_017819339.1 | 5 - 6 | 1110 | 81 | ||
| XM_017963852.1 | XP_017819341.1 | 2 - 3 | 2403 | 81 | ||
More Information
Set Membership: |
|

LDLRAD4
Genomic Map