Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745030.4 | XP_002745076.1 | 5 - 6 | 4140 | 93 | |
| XM_003732356.3 | XP_003732404.1 | 5 - 6 | 2367 | 93 | ||
| XM_008992136.2 | XP_008990384.1 | 5 - 6 | 4139 | 93 | ||
| XM_008992137.2 | XP_008990385.1 | 5 - 6 | 4141 | 93 | ||
| XM_008992139.2 | XP_008990387.1 | 5 - 6 | 4141 | 93 | ||
| XM_008992140.2 | XP_008990388.1 | 5 - 6 | 4140 | 93 | ||
| XM_017969940.1 | XP_017825429.1 | 5 - 6 | 2682 | 93 | ||
| XM_017969942.1 | XP_017825431.1 | 5 - 6 | 1928 | 93 | ||
More Information
Set Membership: |
|

FYB1
Genomic Map