Assay Details
| Gene Symbol: LOC100405150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001649.2 | XP_008999897.1 | 8 - 9 | 1619 | 123 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100405150| Gene Symbol: LOC100405150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001649.2 | XP_008999897.1 | 8 - 9 | 1619 | 123 | |
Set Membership: |
|