Assay Details
| Gene Symbol: GMPR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753669.3 | XP_002753715.1 | 10 - 11 | 1266 | 104 | |
| XM_002753671.3 | XP_002753717.1 | 9 - 10 | 1312 | 104 | ||
| XM_009005822.1 | XP_009004070.1 | 9 - 10 | 1509 | 104 | ||
| XM_009005823.1 | XP_009004071.1 | 9 - 10 | 1434 | 104 | ||
| XM_009005824.1 | XP_009004072.1 | 11 - 12 | 1401 | 104 | ||
| XM_017977027.1 | XP_017832516.1 | 7 - 8 | 1251 | 104 | ||
| GenBank mRNA | EF214848.1 | - | 1 - 2 | 84 | 104 | |
More Information
Set Membership: |
|

GMPR2
Genomic Map