Assay Details
| Gene Symbol: LOC100390305 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756027.2 | XP_002756073.1 | 7 - 8 | 853 | 88 | |
| XM_009009384.2 | XP_009007632.1 | 7 - 8 | 896 | 88 | ||
| XM_017978838.1 | XP_017834327.1 | 7 - 8 | 1449 | 88 | ||
More Information
Set Membership: |
|

LOC100390305
Genomic Map