Assay Details
Gene Symbol: ANG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270922.1 | NP_001257851.1 | 1 | 113 | 130 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001270922.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
