Assay Details
| Gene Symbol: C10H15orf61 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753269.4 | XP_002753315.1 | 1 - 2 | 401 | 92 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Recommended Assay  Probe spans exons | 
 Back To Top
 Back To Top
 C10H15orf61
C10H15orf61 Genomic Map
Genomic Map Back To Top
 Back To Top