Assay Details
| Gene Symbol: IFT27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009010208.2 | XP_009008456.1 | 5 - 6 | 749 | 101 | |
| XM_009010214.2 | XP_009008462.1 | 5 - 6 | 753 | 101 | ||
| XM_017963470.1 | XP_017818959.1 | 5 - 6 | 754 | 101 | ||
More Information
Set Membership: |
|

IFT27
Genomic Map