Assay Details
| Gene Symbol: DRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008996444.2 | XP_008994692.1 | 8 - 9 | 684 | 85 | |
| XM_017972237.1 | XP_017827726.1 | 9 - 10 | 939 | 85 | ||
| XM_017972238.1 | XP_017827727.1 | 9 - 10 | 949 | 85 | ||
More Information
Set Membership: |
|

DRG2
Genomic Map