Assay Details
| Gene Symbol: DHFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986433.2 | XP_008984681.2 | 2 | 314 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008986433.2 transcripts.
Set Membership: |
|

DHFR2
Genomic Map