Assay Details
| Gene Symbol: SULT1A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009009385.2 | XP_009007633.1 | 5 - 6 | 930 | 106 | |
| XM_017978839.1 | XP_017834328.1 | 5 - 6 | 827 | 106 | ||
More Information
Set Membership: |
|

SULT1A3
Genomic Map