Assay Details
| Gene Symbol: PSMC3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747996.4 | XP_002748042.3 | 1 - 2 | 855 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PSMC3IP| Gene Symbol: PSMC3IP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747996.4 | XP_002748042.3 | 1 - 2 | 855 | 71 | |
Set Membership: |
|