Assay Details
| Gene Symbol: RPL26L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744573.4 | XP_002744619.1 | 2 - 3 | 291 | 92 | |
| XM_008990790.1 | XP_008989038.1 | 10 - 11 | 1125 | 92 | ||
| XM_008990794.2 | XP_008989042.1 | 12 - 13 | 1250 | 92 | ||
| XM_008990796.2 | XP_008989044.2 | 11 - 12 | 1281 | 92 | ||
| XM_017969670.1 | XP_017825159.1 | 10 - 11 | 1092 | 92 | ||
| XM_017969673.1 | XP_017825162.1 | 2 - 3 | 353 | 92 | ||
More Information
Set Membership: |
|

RPL26L1
Genomic Map