Assay Details
| Gene Symbol: ATP5PO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986622.2 | XP_008984870.1 | 4 - 5 | 407 | 70 | |
| XM_017967073.1 | XP_017822562.1 | 3 - 4 | 302 | 70 | ||
More Information
Set Membership: |
|

ATP5PO
Genomic Map