Assay Details
| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987455.2 | XP_008985703.1 | 4 - 5 | 480 | 88 | |
| XM_008987456.2 | XP_008985704.1 | 4 - 5 | 480 | 88 | ||
More Information
Set Membership: |
|

CYP4F11
Genomic Map