Assay Details
| Gene Symbol: POLE4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734715.3 | XP_003734763.1 | 1 - 2 | 255 | 101 | |
| XM_008980442.2 | XP_008978690.1 | 1 - 2 | 254 | 101 | ||
| XM_008980443.1 | XP_008978691.1 | 1 - 2 | 297 | 101 | ||
| XM_008980445.2 | XP_008978693.1 | 1 - 2 | 254 | 101 | ||
| XM_008980446.2 | XP_008978694.1 | 1 - 2 | 255 | 101 | ||
| XM_017964219.1 | XP_017819708.1 | 1 - 2 | 254 | 101 | ||
More Information
Set Membership: |
|

POLE4
Genomic Map