Assay Details
| Gene Symbol: BIRC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009006685.2 | XP_009004933.1 | 10 | 3274 | 100 | |
| XM_009006686.2 | XP_009004934.1 | 9 | 3267 | 100 | ||
| XM_009006687.2 | XP_009004935.1 | 10 | 3256 | 100 | ||
| XM_009006688.2 | XP_009004936.1 | 10 | 3091 | 100 | ||
| GenBank mRNA | EF214982.1 | - | 2 | 117 | 100 | |
More Information
Set Membership: |
|

BIRC2
Genomic Map