Assay Details
| Gene Symbol: EDDM3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753740.3 | XP_002753786.1 | 3 | 402 | 106 | |
| XM_009005665.1 | XP_009003913.1 | 2 | 214 | 106 | ||
More Information
Set Membership: |
|

EDDM3A
Genomic Map