Assay Details
| Gene Symbol: INSYN2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744584.3 | XP_002744630.1 | 1 | 231 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002744584.3 transcripts.
Set Membership: |
|

INSYN2B
Genomic Map