Assay Details
| Gene Symbol: UBE2L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806551.4 | XP_002806597.2 | 4 | 2726 | 99 | |
| XM_017978209.1 | XP_017833698.1 | 4 | 2732 | 99 | ||
More Information
Set Membership: |
|

UBE2L3
Genomic Map