Assay Details
| Gene Symbol: PDHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008981903.2 | XP_008980151.1 | 10 | 1162 | 109 | |
| GenBank mRNA | EF215437.1 | - | 1 | 510 | 109 | |
More Information
Set Membership: |
|

PDHB
Genomic Map