Assay Details
| Gene Symbol: AHCY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747265.4 | XP_002747311.1 | 10 | 1938 | 71 | |
| XM_017971825.1 | XP_017827314.1 | 11 | 1927 | 71 | ||
More Information
Set Membership: |
|

AHCY
Genomic Map