Assay Details
| Gene Symbol: NMNAT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008983747.2 | XP_008981995.1 | 5 | 975 | 75 | |
| XM_008983748.2 | XP_008981996.1 | 5 | 972 | 75 | ||
| XM_008983749.1 | XP_008981997.1 | 6 | 885 | 75 | ||
| XM_008983751.2 | XP_008981999.1 | 3 | 710 | 75 | ||
| XM_017965793.1 | XP_017821282.1 | 5 | 784 | 75 | ||
More Information
Set Membership: |
|

NMNAT3
Genomic Map