Assay Details
| Gene Symbol: OAS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009004700.1 | XP_009002948.1 | 2 | 404 | 71 | |
| XM_009004701.2 | XP_009002949.1 | 2 | 393 | 71 | ||
More Information
Set Membership: |
|

OAS1
Genomic Map