Assay Details
| Gene Symbol: LOC100393797 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751152.4 | XP_002751198.1 | 11 | 2473 | 97 | |
| XM_009001795.2 | XP_009000043.1 | 11 | 1573 | 97 | ||
| XM_009001796.2 | XP_009000044.1 | 11 | 1618 | 97 | ||
| XM_009001797.2 | XP_009000045.1 | 11 | 2316 | 97 | ||
| XM_017974983.1 | XP_017830472.1 | 12 | 1670 | 97 | ||
More Information
Set Membership: |
|

LOC100393797
Genomic Map