Assay Details
| Gene Symbol: LOC100415255 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003768.1 | XP_009002016.1 | 1 | 237 | 69 | |
| XM_017976049.1 | XP_017831538.1 | 1 | 237 | 69 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009003768.1, XM_017976049.1 transcripts.
Set Membership: |
|

LOC100415255
Genomic Map