Assay Details
| Gene Symbol: LOC100402578 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762521.3 | XP_002762567.1 | 4 | 1828 | 104 | |
| XM_008988703.2 | XP_008986951.1 | 4 | 1825 | 104 | ||
More Information
Set Membership: |
|

LOC100402578
Genomic Map