Assay Details
Gene Symbol: SPRY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002742596.3 | XP_002742642.1 | 2 | 2268 | 104 | |
XM_002742598.4 | XP_002742644.1 | 2 | 2221 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
