Assay Details
| Gene Symbol: MRPS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984147.2 | XP_008982395.1 | 3 | 591 | 119 | |
| XM_008984148.2 | XP_008982396.1 | 3 | 680 | 119 | ||
More Information
Set Membership: |
|

MRPS17
Genomic Map