Assay Details
| Gene Symbol: SDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753040.4 | XP_002753086.2 | 8 | 1019 | 75 | |
| GenBank mRNA | EF215079.1 | - | 1 | 125 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SDS| Gene Symbol: SDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753040.4 | XP_002753086.2 | 8 | 1019 | 75 | |
| GenBank mRNA | EF215079.1 | - | 1 | 125 | 75 | |
Set Membership: |
|