Assay Details
| Gene Symbol: LOC100411160 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987451.2 | XP_008985699.1 | 11 | 3834 | 89 | |
| XM_008987453.2 | XP_008985701.2 | 12 | 2520 | 89 | ||
| XM_017967878.1 | XP_017823367.1 | 12 | 3749 | 89 | ||
More Information
Set Membership: |
|

LOC100411160
Genomic Map