Assay Details
| Gene Symbol: ACTR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757688.4 | XP_002757734.1 | 10 | 2967 | 121 | |
| XM_002757689.4 | XP_002757735.1 | 9 | 2952 | 121 | ||
| XM_008980602.2 | XP_008978850.1 | 10 | 2559 | 121 | ||
More Information
Set Membership: |
|

ACTR2
Genomic Map