Assay Details
| Gene Symbol: LOC100401402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752307.4 | XP_002752353.3 | 2335 | 109 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100401402| Gene Symbol: LOC100401402 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752307.4 | XP_002752353.3 | 2335 | 109 | ||