Assay Details
| Gene Symbol: CMPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750795.3 | XP_002750841.1 | 6 | 1215 | 101 | |
| XM_009001190.1 | XP_008999438.1 | 5 | 1068 | 101 | ||
More Information
Set Membership: |
|

CMPK1
Genomic Map