Assay Details
| Gene Symbol: STON1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204840.1 | NP_001191769.1 | 1 - 2 | 1967 | 110 | |
| XM_008980717.1 | XP_008978965.1 | 2 - 3 | 2044 | 110 | ||
| XM_008980718.2 | XP_008978966.1 | 2 - 3 | 2720 | 110 | ||
More Information
Set Membership: |
|

STON1
Genomic Map